missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXER1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Especificaciones
Antígeno | OXER1 |
---|---|
Concentración | 0.25 mg/mL |
Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry |
Clasificación | Polyclonal |
Conjugado | Unconjugated |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16826695
|
Proteintech
55202-1-AP-20UL |
20 μL |
140.00€
20 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
16816695
|
Proteintech
55202-1-AP-150UL |
150 μL |
399.00€
150 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
This antibody is specific to OXER1.
TG1019 is a G protein-coupled receptor that functions as a chemoattractant for eosinophils and neutrophils. TG1019 is expressed in various tissues except the brain, with the highest expression in liver, kidney, peripheral leukocyte, lung and spleen. TG1019 interacts with the protein OXE to form TG1019/OXE, a Galpha i/o protein-coupled receptor. Signals from TG1019 are transduced via a Galpha i/o protein to PLC/calcium, MEK/ERK and PI3K/Akt pathways. Signal transduction from TG1019 following stimulation with 5-oxo-6E,8Z,11Z,14Z eicosatetraenoic acid (5-oxo-ETE) induces intracellular calcium mobilization and rapid activation of MEK/ERK and PI3K/Akt pathways. TG1019 also may play a role in downregulating cAMP production.Especificaciones
OXER1 | |
Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human | |
Q8TDS5 | |
165140 | |
Peptide | |
Primary | |
-20°C | |
OXER1 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
G protein coupled receptor 170, GPCR, GPR170, OXER1, oxoeicosanoid (OXE) receptor 1, Oxoeicosanoid receptor 1, TG1019 | |
OXER1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |