missing translation for 'onlineSavingsMsg'
Learn More

OSBPL9 Antibody, Novus Biologicals™

Código de producto. 18652514 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
Código de producto. Cantidad unitSize
18652514 50 μg 50 microlitros
18674782 100 μg 100 microlitros
2 options
This item is not returnable. View return policy

Product Code. 18652514

Brand: Novus Biologicals NBP29724550ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OSBPL9 Polyclonal antibody specifically detects OSBPL9 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno OSBPL9
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunohistochemistry-Paraffin 1:200-1:1000
Formulación PBS (pH 7.0)
Alias de gen FLJ12492, FLJ14629, FLJ14801, FLJ32055, FLJ34384, ORP-9, ORP9MGC15035, OSBP4, OSBP-related protein 9, oxysterol binding protein-like 9, oxysterol-binding protein-related protein 9
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with E. coli-derived Human OSBPL9 fragment.
Método de purificación Antigen and protein A Affinity-purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Lipid and Metabolism
Primario o secundario Primary
ID de gen (Entrez) 114883
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.