missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
OSBPL11 Monoclonal specifically detects OSBPL11 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Especificaciones
Especificaciones
| Antígeno | OSBPL11 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | OTI6A6 |
| Conjugado | PE |
| Dilución | Flow Cytometry |
| Formulación | PBS with 0.05% Sodium Azide |
| Alias de gen | FLJ13012, FLJ13164, ORP11OSBP-related protein 11, ORP-11TCCCIA00292, OSBP12, oxysterol binding protein-like 11, oxysterol-binding protein-related protein 11 |
| Símbolos de los genes | OSBPL11 |
| Especie del huésped | Mouse |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?