missing translation for 'onlineSavingsMsg'
Learn More

ORC6L Antibody, DyLight 594, Novus Biologicals™

Código de producto. 18764211 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18764211 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18764211 Proveedor Novus Biologicals N.º de proveedor NB10074619DL594

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

ORC6L Polyclonal antibody specifically detects ORC6L in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno ORC6L
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado DyLight 594
Dilución Western Blot
Formulación 50mM Sodium Borate
Alias de gen ORC6Lorigin recognition complex, subunit 6 homolog-like (yeast), origin recognition complex subunit 6, origin recognition complex, subunit 6, origin recognition complex, subunit 6 like (yeast)
Especie del huésped Rabbit
Inmunógeno The immunogen recognized by this antibody maps to a region between residue 200 and 230 of human origin recognition complex subunit 6 using the numbering given in entry NP_055136.1 (GeneID 23594).
Método de purificación Immunogen affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación DNA replication Transcription Translation and Splicing
Primario o secundario Primary
ID de gen (Entrez) 23594
Especies diana Human
Contenido y almacenamiento Store at 4C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.