missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Oncostatin M/OSM Monoclonal specifically detects Oncostatin M/OSM in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
| Antígeno | Oncostatin M/OSM |
| Aplicaciones | Western Blot, Immunohistochemistry |
| Clasificación | Monoclonal |
| Clon | OTI2B6 |
| Conjugado | DyLight 650 |
| Dilución | Western Blot, Immunohistochemistry |
| Formulación | 50mM Sodium Borate with 0.05% Sodium Azide |
| Alias de gen | MGC20461, oncostatin M, oncostatin-M |
| Símbolos de los genes | OSM |
| Especie del huésped | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?