missing translation for 'onlineSavingsMsg'
Learn More

OCT4 Antibody, Novus Biologicals™

Código de producto. 18232935 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18232935 50 μg 50 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18232935 Proveedor Novus Biologicals N.º de proveedor NBP259164

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

44473 Polyclonal specifically detects 44473 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Chromatin Immunoprecipitation Sequencing.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno OCT4
Aplicaciones Western Blot, ELISA, ChIP Assay
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:1000, ELISA 1:1000-1:10000, Chromatin Immunoprecipitation (ChIP) 5 ug/IP, Chromatin Immunoprecipitation Sequencing
Formulación PBS with 0.05% Sodium Azide and 0.05% ProClin 300
Alias de gen class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein
Símbolos de los genes POU5F1
Especie del huésped Rabbit
Inmunógeno The exact immunogen is propietary information.
Método de purificación Affinity Purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Cellular Markers, Chromatin Research, Embryonic Stem Cell Markers, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators
Primario o secundario Primary
ID de gen (Entrez) 5460
Especies diana Human
Contenido y almacenamiento Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.