missing translation for 'onlineSavingsMsg'
Learn More

OCT4 Antibody (C7), Novus Biologicals™

Código de producto. 30279222 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
30279222 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. View return policy

Product Code. 30279222

Brand: Novus Biologicals NBP343284

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

OCT4 Monoclonal antibody specifically detects OCT4 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antígeno OCT4
Aplicaciones Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
Clasificación Monoclonal
Clon C7
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 0.5-2ug/mL, Immunohistochemistry 5-20 μg/mL, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunoprecipitation
Formulación 0.01 M PBS, pH 7.4, 50% glycerol
Alias de gen class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein
Especie del huésped Mouse
Inmunógeno Immunized with recombinant Human OCT4 (Uniprot#: Q01860; Ser136∽Asn360)
Método de purificación Protein A or G purified
Cantidad 100 μL
Estado normativo RUO
Disciplina de investigación Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators
Primario o secundario Primary
ID de gen (Entrez) 5460
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.