missing translation for 'onlineSavingsMsg'
Learn More

OCT2 Antibody, Novus Biologicals™

Código de producto. 18680318 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
25 μL
Tamaño de la unidad:
0.10 ml
25 microlitros
Código de producto. Cantidad unitSize
18680318 25 μL 25 microlitros
18677397 0.1 mL 0.10 ml
2 options
This item is not returnable. View return policy

Product Code. 18680318

Brand: Novus Biologicals NBP24941825ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OCT2 Polyclonal antibody specifically detects OCT2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno OCT2
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000
Formulación PBS (pH 7.2), 40% Glycerol
Alias de gen homeobox protein, Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2, OCT2Oct-2, Octamer-binding protein 2, Octamer-binding transcription factor 2, OTF-2, OTF2POU domain class 2, transcription factor 2, POU class 2 homeobox 2, POU domain, class 2, transcription factor 2
Especie del huésped Rabbit
Inmunógeno This antibody was developed against a recombinant protein corresponding to amino acids: PAQFLLPQAQQSQPGLLPTPNLFQLPQQTQGALLTSQPRAGLPTQAVTRPTLPDPHLSHPQPPKCLEPPSH
Método de purificación Immunogen affinity purified
Cantidad 25 μL
Estado normativo RUO
Disciplina de investigación Cancer
Primario o secundario Primary
ID de gen (Entrez) 5452
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.