missing translation for 'onlineSavingsMsg'
Learn More

NVP-QAV-572, MedChemExpress

Código de producto. 16437096
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
16437096 100 mg 100 miligramos
16483087 50 mg 50 miligramos
16477038 10 mg 10 miligramos
16429587 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16437096 Supplier Medchem Express Supplier No. HY16355

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress NVP-QAV-572 is a PI3K inhibitor extracted from patent US7998990B2, Compound Example 8, has an IC50 of 10 nM.

NVP-QAV-572 is a PI3K inhibitor extracted from patent US7998990B2, Compound Example 8, has an IC50 of 10 nM.

Specifications

Nombre del producto químico o material NVP-QAV-572
CAS 957209-68-6
Color Blanco
Notas de grado de pureza Research
Fórmula molecular C17H19F2N7O3S2
Cantidad 100 mg
Información de solubilidad DMSO : 125 mg/mL (265.11 mM; Need ultrasonic)
SMILES O=C(NC1=NC(C)=C(C2=CC=C(S(=O)(C)=O)C(F)=C2)S1)NCCC3=NN(CCF)N=N3
Peso molecular (g/mol) 471.5
Formula Weight (peso de la fórmula) 471.5
Porcentaje de pureza 98.05%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.