missing translation for 'onlineSavingsMsg'
Learn More

Nucleolin Antibody (NCL/902) - IHC-Prediluted, Novus Biologicals™

Código de producto. 18100158 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
7 mL
Tamaño de la unidad:
7 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18100158 7 mL 7 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18100158 Proveedor Novus Biologicals N.º de proveedor NBP248317

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Nucleolin Monoclonal specifically detects Nucleolin in Human, Bovine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Nucleolin
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon NCL/902
Conjugado Unconjugated
Dilución Immunohistochemistry, Immunohistochemistry-Paraffin
Formulación 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Alias de gen C23, FLJ45706, FLJ59041, nucleolin, Protein C23
Símbolos de los genes NCL
Especie del huésped Mouse
Inmunógeno Recombinant full-length human Nucleolin protein (Uniprot: P19338)
Peso molecular del antígeno 76 kDa
Método de purificación Protein A or G purified
Cantidad 7 mL
Estado normativo RUO
Disciplina de investigación Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 4691
Especificidad de la prueba Recognizes a protein of ∼76kDa, which is identified as Nucleolin (NCL). It is the major nucleolar phosphoprotein of growing eukaryotic cells. NCL is located mainly in dense fibrillar regions of the nucleolus. It is found associated with intranucleolar chromatin and pre-ribosomal particles. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. It induces chromatin decondensation by binding to histone H1. It is thought to play a role in pre-rRNA transcription and ribosome assembly. This Mouse Monoclonal Antibody can be used to stain the nucleoli in cell or tissue preparations and can be used as a marker of the nucleoli in subcellular fractions. It produces a speckled pattern in the nuclei of cells of normal and malignant cells and may be used to stain the nucleoli of cells in fixed or frozen tissue sections. It can be used with paraformaldehyde fixed frozen tissue or cell preparations and formalin fixed, paraffin-embedded tissue sections.
Especies diana Human, Bovine (Negative), Mouse (Negative), Rat (Negative)
Contenido y almacenamiento Store at 4C.
Formulario Purified
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.