missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
NSUN4 Polyclonal antibody specifically detects NSUN4 in Human, Mouse, Rat samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | NSUN4 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000 |
| Formulación | PBS (pH 7.3), 50% glycerol |
| Alias de gen | EC 2.1.1, EC 2.1.1.-, EC 2.1.1.29, MGC22960, NOL1/NOP2/Sun domain family 4, NOL1/NOP2/Sun domain family member 4, NOL1/NOP2/Sun domain family, member 4, NOP2/Sun domain family, member 4, putative methyltransferase NSUN4, RP4-603I14.2 |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fusion protein containing a sequence corresponding to amino acids 26-290 of human NSUN4 (NP_950245.2). RYKKKWAATEPKFPAVRLALQNFDMTYSVQFGDLWPSIRVSLLSEQKYGALVNNFAAWDHVSAKLEQLSAKDFVNEAISHWELQSEGGQSAAPSPASWACSPNLRCFTFDRGDISRFPPARPGSLGVMEYYLMDAASLLPVLALGLQPGDIVLDLCAAPGGKTLALLQTGCCRNLAANDLSPSRIARLQKILHSYVPEEIRDGNQVRVTSWDGRKWGELEGDTYDRVLVDVPCTTDRHSLHEEENNIFKRSRKKERQILPVLQVQ |
| Método de purificación | Affinity purified |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?