Descripción
NSUN4 Polyclonal antibody specifically detects NSUN4 in Human, Mouse, Rat samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | NSUN4 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000 |
| Formulación | PBS (pH 7.3), 50% glycerol |
| Alias de gen | EC 2.1.1, EC 2.1.1.-, EC 2.1.1.29, MGC22960, NOL1/NOP2/Sun domain family 4, NOL1/NOP2/Sun domain family member 4, NOL1/NOP2/Sun domain family, member 4, NOP2/Sun domain family, member 4, putative methyltransferase NSUN4, RP4-603I14.2 |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fusion protein containing a sequence corresponding to amino acids 26-290 of human NSUN4 (NP_950245.2). RYKKKWAATEPKFPAVRLALQNFDMTYSVQFGDLWPSIRVSLLSEQKYGALVNNFAAWDHVSAKLEQLSAKDFVNEAISHWELQSEGGQSAAPSPASWACSPNLRCFTFDRGDISRFPPARPGSLGVMEYYLMDAASLLPVLALGLQPGDIVLDLCAAPGGKTLALLQTGCCRNLAANDLSPSRIARLQKILHSYVPEEIRDGNQVRVTSWDGRKWGELEGDTYDRVLVDVPCTTDRHSLHEEENNIFKRSRKKERQILPVLQVQ |
| Método de purificación | Affinity purified |
| Mostrar más |