missing translation for 'onlineSavingsMsg'
Learn More

Noxa Antibody (S07-3H6), Novus Biologicals™

Código de producto. 18636653 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18636653 100 μg 100 microlitros
18605763 50 μg 50 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18636653 Proveedor Novus Biologicals N.º de proveedor NBP319820100ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Monoclonal Antibody

Noxa Monoclonal antibody specifically detects Noxa in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Noxa
Aplicaciones Western Blot
Clasificación Monoclonal
Clon S07-3H6
Concentración 0.3 mg/mL
Conjugado Unconjugated
Formulación 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Alias de gen adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa
Especie del huésped Rabbit
Inmunógeno A synthetic peptide of human Noxa (Uniprot # Q13794)
Método de purificación Affinity purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Apoptosis, Cancer, Cellular Markers, Lipid and Metabolism, Membrane Trafficking and Chaperones, Mitochondrial Mediated Pathway, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors
Primario o secundario Primary
ID de gen (Entrez) 5366
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.