Learn More
NOX1 Antibody - BSA Free, Novus Biologicals™
Código de producto. 18676722 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.02 mL
0.1 mL
Tamaño de la unidad:
0.01 ml
0.02 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18676722 0.1 mL 0.01 ml
18663002 0.02 mL 0.02 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18676722 Proveedor Novus Biologicals N.º de proveedor NBP2947520.1ml
Rabbit Polyclonal Antibody
NOX1 Polyclonal antibody specifically detects NOX1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno NOX1
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:100 - 1:500
Formulación PBS (pH 7.3), 50% glycerol
Alias de gen EC 1.6.3, GP91-2, Mitogenic oxidase 1, MOX-1, MOX1NADH/NADPH mitogenic oxidase subunit P65-MOX, NADPH oxidase 1, NADPH oxidase homolog-1, NOH1mitogenic oxidase (pyridine nucleotide-dependent superoxide-generating), NOH-1NADPH oxidase 1 variant NOH-1L, NOX-1
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 415-564 of human NOX1 (NP_008983.2). KSIWYKFQCADHNLKTKKIYFYWICRETGAFSWFNNLLTSLEQEMEELGKVGFLNYRLFLTGWDSNIVGHAALNFDKATDIVTGLKQKTSFGRPMWDNEFSTIATSHPKSVVGVFLCGPRTLAKSLRKCCHRYSSLDPRKVQFYFNKENF
Método de purificación Affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cancer, Cell Biology, Direct Reversal of DNA Damage, Endocrinology, Immunology, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 27035
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG