missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDA R, NR1 Subunit Antibody (1031915), Alexa Fluor™ 488, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
NMDA R, NR1 Subunit Monoclonal antibody specifically detects NMDA R, NR1 Subunit in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry
Especificaciones
Especificaciones
| Antígeno | NMDA R, NR1 Subunit |
| Aplicaciones | Flow Cytometry, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 1031915 |
| Conjugado | Alexa Fluor 488 |
| Dilución | Intracellular Staining by Flow Cytometry, Immunocytochemistry |
| Formulación | 50mM Sodium Borate |
| Alias de gen | GluN1, MRD8, NDHMSD, NDHMSR,NMD-R1, NMDA1, NMDAR1, NR1 |
| Especie del huésped | Rat |
| Inmunógeno | Mouse myeloma cell line NS0-derived recombinant mouse GRIN1/NMDAR1, Met1-Gln559 |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?