Learn More
NLRP7 Antibody, Biotin, Novus Biologicals™
Código de producto. 18570111 Tienda Bio Techne Productos
Change view
Click to view available options
:
0.1 mL; Biotin
Código de producto.
18570111 0.1 mL; Biotin
1 options
Código de producto. 18570111
Marca: Novus Biologicals NBP176290B
Rabbit Polyclonal Antibody
NLRP7 Polyclonal antibody specifically detects NLRP7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno NLRP7
Aplicaciones Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Biotin
Formulación PBS
Alias de gen CLR19.4, MGC126471, NACHT, leucine rich repeat and PYD containing 7, NACHT, LRR and PYD containing protein 7, NACHT, LRR and PYD domains-containing protein 7, NALP7MGC126470, NLR family, pyrin domain containing 7, NOD12HYDM, Nucleotide-binding oligomerization domain protein 12, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 7, PAN7, PYPAF3FLJ94610, PYRIN-containing APAF1-like protein 3
Especie del huésped Rabbit
Inmunógeno Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human NALP7. The immunogen is located within amino acids 90 - 140 of NALP7. Amino Acid Squence: LGDAEEDSELAKPGEKEG
Método de purificación Peptide affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Apoptosis, Cell Biology, Immunology, Innate Immunity
Primario o secundario Primary
ID de gen (Entrez) 199713
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG