missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Nitrilase 1 Monoclonal specifically detects Nitrilase 1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Especificaciones
Especificaciones
| Antígeno | Nitrilase 1 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | OTI3A11 |
| Conjugado | APC |
| Dilución | Flow Cytometry |
| Formulación | PBS with 0.05% Sodium Azide |
| Alias de gen | EC 3.5, MGC57670, nitrilase 1, nitrilase homolog 1 |
| Símbolos de los genes | NIT1 |
| Especie del huésped | Mouse |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?