missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
NIRF Polyclonal antibody specifically detects NIRF in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | NIRF |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 647 |
| Formulación | 50 mM Sodium Borate |
| Alias de gen | DKFZp434B0920, DKFZp686G0837, E3 ubiquitin-protein ligase UHRF2, EC 6.3.2, EC 6.3.2.-, MGC33463, NIRFUbiquitin-like-containing PHD and RING finger domains protein 2, Np95/ICBP90-like RING finger protein, Np95-like RING finger protein, Nuclear protein 97, Nuclear zinc finger protein Np97, RNF107RING finger protein 107, Ubiquitin-like PHD and RING finger domain-containing protein 2, ubiquitin-like with PHD and ring finger domains 2, ubiquitin-like, containing PHD and RING finger domains, 2, URF2 |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NIRF. |
| Método de purificación | Antigen and protein A Affinity-purified |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?