Learn More
NHERF-1 Antibody (EBP-10), Alexa Fluor™ 594, Novus Biologicals™
Código de producto. 18241132 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
18241132 0.1 mL 0.10 ml
1 options
Código de producto. 18241132
Marca: Novus Biologicals NB500488AF594
Mouse Monoclonal Antibody
NHERF-1 Monoclonal specifically detects NHERF-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno NHERF-1
Aplicaciones Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon EBP-10
Conjugado Alexa Fluor 594
Dilución Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulación 50 mM sodium borate with 0.05% sodium azide
Alias de gen EBP50NHERF-1, ezrin-radixin-moesin binding phosphoprotein-50, Ezrin-radixin-moesin-binding phosphoprotein 50, Na(+)/H(+) exchange regulatory cofactor NHE-RF1, Na+/H+ exchange regulatory co-factor, NHERF1solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 1, NHERFNPHLOP2, Regulatory cofactor of Na(+)/H(+) exchanger, sodium/hydrogen exchanger regulatory factor 1, Sodium-hydrogen exchanger regulatory factor 1, solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatoryfactor 1, solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 1, Solute carrier family 9 isoform A3 regulatory factor 1
Símbolos de los genes SLC9A3R1
Especie del huésped Mouse
Inmunógeno Bacterially produced recombinant full-length human NHERF1.
Cantidad 0.1 mL
Primario o secundario Primary
ID de gen (Entrez) 9368
Especificidad de la prueba The antibody EBP-10 reacts with NHERF1/EBP50 phosphoprotein of 50 kDa, which serves as an adaptor and regulator protein.
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Isotype IgG2b