missing translation for 'onlineSavingsMsg'
Learn More

NF-L Antibody (NFL/736), PE, Novus Biologicals™

Código de producto. 18614088 Tienda Bio Techne Productos
Change view
Click to view available options
:
0.1 mL, PE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto.
18614088 0.1 mL, PE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18614088 Proveedor Novus Biologicals N.º de proveedor NBP247970PE

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

NF-L Monoclonal antibody specifically detects NF-L in Human, Rat, Porcine, Bovine, Chicken samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antígeno NF-L
Aplicaciones Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Clasificación Monoclonal
Clon NFL/736
Conjugado PE
Formulación PBS
Alias de gen 68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa
Especie del huésped Mouse
Inmunógeno Recombinant human NF-L protein (Uniprot: P07196)
Método de purificación Protein A or G purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primario o secundario Primary
ID de gen (Entrez) 4747
Especies diana Human, Rat, Pig, Bovine, Chicken
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.