missing translation for 'onlineSavingsMsg'
Learn More

NF-H Antibody (NEFL.H/2324R), Janelia Fluor™ 549, Novus Biologicals™

Código de producto. 18587783 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
18587783 100 μg 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18587783

Marca: Novus Biologicals NBP308552JF549

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

NF-H Monoclonal antibody specifically detects NF-H in Human, Mouse, Rat, Porcine, Chicken samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno NF-H
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon NEFL.H/2324R
Conjugado Janelia Fluor 549
Formulación 50mM Sodium Borate
Alias de gen 200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H
Especie del huésped Rabbit
Inmunógeno Recombinant full-length human NF-H protein (Uniprot: P12036)
Método de purificación Protein A or G purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primario o secundario Primary
ID de gen (Entrez) 4744
Especies diana Human, Mouse, Rat, Pig, Chicken
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.