Learn More
NF-H Antibody (CL2671) [DyLight 755], Novus Biologicals™
Código de producto. 30295407 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30295407 0.1 mL 0.10 ml
1 options
Código de producto. 30295407
Marca: Novus Biologicals NBP246615IR
Mouse Monoclonal Antibody
NF-H Monoclonal antibody specifically detects NF-H in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno NF-H
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CL2671
Conjugado DyLight 755
Formulación 50mM Sodium Borate
Alias de gen 200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from P12036, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primario o secundario Primary
ID de gen (Entrez) 4744
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1