missing translation for 'onlineSavingsMsg'
Learn More

Neuropilin-1 Antibody (761704) [PE], Novus Biologicals™

Código de producto. 18617214 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18617214 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Product Code. 18617214 Supplier Novus Biologicals Supplier No. FAB59941P

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rat Monoclonal Antibody

Neuropilin-1 Monoclonal antibody specifically detects Neuropilin-1 in Mouse samples. It is validated for Flow Cytometry, Neutralization, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Neuropilin-1
Aplicaciones Flow Cytometry, Neutralization, CyTOF
Clasificación Monoclonal
Clon 761704
Conjugado PE
Dilución Flow Cytometry, Neutralization, CyTOF-ready
Formulación PBS
Alias de gen BDCA4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, NP1, NRP, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R
Especie del huésped Rat
Inmunógeno Mouse myeloma cell line NS0-derived recombinant mouse Neuropilin-1, Phe21-Pro856, Accession # P97333
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Adaptive Immunity, Angiogenesis, Breast Cancer, Cancer, Cardiovascular Biology, Neuroscience, Virology Bacteria and Parasites
Primario o secundario Primary
ID de gen (Entrez) 8829
Especies diana Mouse
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.