missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Neuroligin 3/NLGN3 Monoclonal specifically detects Neuroligin 3/NLGN3 in Human samples. It is validated for Flow Cytometry.
Especificaciones
Especificaciones
| Antígeno | Neuroligin 3/NLGN3 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 242 |
| Conjugado | Unconjugated |
| Dilución | Flow Cytometry 1:25-1:100 |
| Formulación | 0.2 um filtered solution in PBS |
| Alias de gen | ASPGX1, AUTSX1, Gliotactin homolog, KIAA1480HNL3, neuroligin 3, neuroligin-3, NL3 |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant Human Neuroligin 3/NLGN3 Protein |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?