missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Neurogenin-3 Antibody (NGN3/1809) - Azide and BSA Free, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Neurogenin-3 Monoclonal antibody specifically detects Neurogenin-3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Especificaciones
Specifica
| Antígeno | Neurogenin-3 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | NGN3/1809 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Formulación | 10 mM PBS |
| Alias de gen | Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, neurogenin 3, neurogenin-3, NGN3, Protein atonal homolog 5 |
| Especie del huésped | Mouse |
| Vedi altri risultati |
Titolo del prodotto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?