missing translation for 'onlineSavingsMsg'
Learn More

Neurofibromin 1 Antibody (McNFn27b), DyLight 594, Novus Biologicals™

Código de producto. 18794210 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.2 mL
Tamaño de la unidad:
0.20 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18794210 0.2 mL 0.20 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18794210 Proveedor Novus Biologicals N.º de proveedor NB300154DL594

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Neurofibromin 1 Monoclonal antibody specifically detects Neurofibromin 1 in Human, Mouse, Rat, Guinea Pig samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Neurofibromin 1
Aplicaciones ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon McNFn27b
Conjugado DyLight 594
Dilución ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulación 50mM Sodium Borate
Alias de gen DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS
Especie del huésped Mouse
Inmunógeno A highly conserved N-terminal peptide, corresponding to amino acids 27-41 of human Neurofibromin 1. [UniProt# P21359]
Método de purificación Protein A or G purified
Cantidad 0.2 mL
Estado normativo RUO
Disciplina de investigación Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors
Primario o secundario Primary
ID de gen (Entrez) 4763
Especies diana Human, Mouse, Rat, Guinea Pig
Contenido y almacenamiento Store at 4C in the dark.
Formulario Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.