Learn More
Nestin Antibody (CL0197) [mFluor Violet 610 SE], Novus Biologicals™
Código de producto. 30291872 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30291872 0.1 mL 0.10 ml
1 options
Código de producto. 30291872
Marca: Novus Biologicals NBP259775MFV610
Mouse Monoclonal Antibody
Nestin Monoclonal antibody specifically detects Nestin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Nestin
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
Clasificación Monoclonal
Clon CL0197
Conjugado mFluor Violet 610 SE
Formulación 50mM Sodium Borate
Alias de gen FLJ21841, nestin
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from P48681, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Angiogenesis, Cancer, Cancer Stem Cells, Cell Biology, Cellular Markers, Extracellular Matrix, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 10763
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1