Learn More
Nesprin 2 Antibody (6C11) - BSA Free, Novus Biologicals™
Código de producto. 18634508 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.1 mg
Tamaño de la unidad:
0.03 miligramo
0.10 miligramo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18634508 0.025 mg 0.03 miligramo
18685328 0.1 mg 0.10 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18634508 Proveedor Novus Biologicals N.º de proveedor NBP2599440.025mg
Mouse Monoclonal Antibody
Nesprin 2 Monoclonal antibody specifically detects Nesprin 2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Nesprin 2
Aplicaciones Western Blot, Immunofluorescence
Clasificación Monoclonal
Clon 6C11
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:100 - 1-2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Formulación PBS
Alias de gen DKFZP434H2235, DKFZp686E01115, DKFZp686H1931, EDMD5, FLJ46790, KIAA1011FLJ11014, nesprin 2, nesprin-2, NUAFLJ43727, NUANCEFLJ45710, Nuclear envelope spectrin repeat protein 2, Nucleus and actin connecting element protein, polytrophin, Protein NUANCE, spectrin repeat containing, nuclear envelope 2, Synaptic nuclear envelope protein 2, synaptic nuclei expressed gene 2, SYNE-2, TROPH
Especie del huésped Mouse
Inmunógeno GST fused to residues 6,653-6,835 of human Nesprin 2
Método de purificación Protein A purified
Cantidad 0.025 mg
Estado normativo RUO
Disciplina de investigación Developmental Biology
Primario o secundario Primary
ID de gen (Entrez) 23224
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG2b κ