Learn More
Eurofins DiscoverX NEK11 Protein, Active
Código de producto. 17360584
Encompass_Preferred
Change view
Click to view available options
Cantidad:
10 μg
250 μg
Tamaño de la unidad:
10 microgramos
250 microgramos
17360584 10 μg 10 microgramos
17370584 250 μg 250 microgramos
2 options
Código de producto. 17360584
Marca: Eurofins DiscoverX 14700
Active, 6His-tagged, recombinant, human full length, NEK11, for use in Kinase Assays.
Protein kinase which plays an important role in the G2/M checkpoint response to DNA damage. Controls degradation of CDC25A by directly phosphorylating it on residues whose phosphorylation is required for BTRC-mediated polyubiquitination and degradation.
GenBank BC028587
Especificaciones
Formulación mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Peso molecular 59.3 kDa
Cantidad 10 μg
Especie Human
Recombinante Recombinant
Etiqueta de proteína N-terminal 6His
Sistema de expresión Recombinant enzyme expressed in Sf21 insect cells
Forma de proteína Full-length
Grado de pureza o calidad ≥61%
Expresión NEK11, active