Learn More
NEDD4 Antibody (S06-7C3), Novus Biologicals™
Código de producto. 18648493 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
18648493 100 μg 100 microlitros
18657663 50 μg 50 microlitros
2 options
Código de producto. 18648493
Marca: Novus Biologicals NBP319815100ul
Rabbit Monoclonal Antibody
NEDD4 Monoclonal antibody specifically detects NEDD4 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno NEDD4
Aplicaciones Western Blot
Clasificación Monoclonal
Clon S06-7C3
Concentración 0.3 mg/mL
Conjugado Unconjugated
Formulación 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Alias de gen Cell proliferation-inducing gene 53 protein, E3 ubiquitin-protein ligase NEDD4, EC 6.3.2, EC 6.3.2.-, KIAA0093NEDD4-1MGC176705, NEDD-4, Neural precursor cell expressed developmentally down-regulated protein 4, neural precursor cell expressed, developmentally down-regulated 4, receptor-potentiating factor 1, RPF1
Especie del huésped Rabbit
Inmunógeno A synthetic peptide of human NEDD4 (Uniprot # P46934)
Método de purificación Affinity purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Cell Biology, Cell Cycle and Replication, Endocrinology, Immunology, Neuroscience, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 4734
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG