Learn More
Novus Biologicals™ Nav1.7 Antibody (Duke anti-NAv1.7) - Humanized, Novus Biologicals™
Código de producto. 30044922 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
30044922 1 mg 1 miligramo
30047229 100 μg 100 microgramos
30045768 50 μg 50 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 30044922 Proveedor Novus Biologicals™ N.º de proveedor NBP3279871mg
Human Monoclonal Antibody
Nav1.7 Monoclonal antibody specifically detects Nav1.7 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Nav1.7
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon Duke anti-NAv1.7
Conjugado Unconjugated
Formulación Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
N.º de referencia del gen Q15858
Alias de gen ETHA, FEB3B, NE-NA, NENAVoltage-gated sodium channel subunit alpha Nav1.7, Neuroendocrine sodium channel, Peripheral sodium channel 1, PN1hNE-Na, sodium channel protein type 9 subunit alpha, Sodium channel protein type IX subunit alpha, sodium channel, voltage-gated, type IX, alpha subunit, voltage-gated sodium channel alpha subunit Nav1.7, voltage-gated, type IX, alpha polypeptide
Especie del huésped Human
Inmunógeno SCN9a/ Nav1.7
Método de purificación Protein A purified
Cantidad 1 mg
Estado normativo RUO
Disciplina de investigación Cancer, Cellular Signaling, Neuroscience
Primario o secundario Primary
ID de gen (Entrez) 6335
Reconstitución Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Especies diana Human
Contenido y almacenamiento Store at -20°C in powder form. Store at -80°C once reconstituted.
Formulario Purified
Isotype IgG1