Learn More
N-Cadherin / Cadherin-2 / CD325 Mouse, Clone: CDH2/1573, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 1000-MSM3-P0
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| N-Cadherin / Cadherin-2 / CD325 | |
| Cell surface | |
| CDH2/1573 | |
| Unconjugated | |
| NP_001295105 | |
| Mouse | |
| 130-140kDa | |
| Protein A/G Purified | |
| RUO | |
| 1000 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| CDH2 | |
| Recombinant human CDH2 intracellular domain | |
| HeLa or HUVEC cells. Heart, Pancreas or Cerebral Cortex | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |