Learn More
N-Cadherin Antibody (CL3716) [mFluor Violet 610 SE], Novus Biologicals™
Código de producto. 30304032 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30304032 0.1 mL 0.10 ml
1 options
Código de producto. 30304032
Marca: Novus Biologicals NBP261632MFV610
Mouse Monoclonal Antibody
N-Cadherin Monoclonal antibody specifically detects N-Cadherin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno N-Cadherin
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
Clasificación Monoclonal
Clon CL3716
Conjugado mFluor Violet 610 SE
Formulación 50mM Sodium Borate
Alias de gen ACOGS, ARVD14, cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, calcium-dependent adhesion protein, neuronal, CD325, CD325 antigen, CDH2, CDHN, CDw325, NCAD, N-cadherin, N-cadherin 1, Neural cadherin, neural-cadherin
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from P19022, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells
Primario o secundario Primary
ID de gen (Entrez) 1000
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1