missing translation for 'onlineSavingsMsg'
Learn More

N-Cadherin Antibody (CDH2/8862R), Novus Biologicals™

Código de producto. 18622435 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
20 μg
Tamaño de la unidad:
100 microgramos
20 microgramos
Código de producto. Cantidad unitSize
18622435 20 μg 20 microgramos
18632464 100 μg 100 microgramos
2 options
This item is not returnable. View return policy

Product Code. 18622435

Brand: Novus Biologicals NBP32384520ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

N-Cadherin Monoclonal antibody specifically detects N-Cadherin in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno N-Cadherin
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CDH2/8862R
Concentración 0.2 mg/mL
Conjugado Unconjugated
Dilución Immunohistochemistry-Paraffin : 1-2 μg/mL
Formulación 10 mM PBS with 0.05% BSA
Alias de gen ACOGS, ARVD14, cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, calcium-dependent adhesion protein, neuronal, CD325, CD325 antigen, CDH2, CDHN, CDw325, NCAD, N-cadherin, N-cadherin 1, Neural cadherin, neural-cadherin
Especie del huésped Rabbit
Inmunógeno Recombinant fragment (around aa605-905) of human N-Cadherin protein (exact sequence is proprietary)
Método de purificación Protein A or G purified
Cantidad 20 μg
Estado normativo RUO
Disciplina de investigación Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells
Primario o secundario Primary
ID de gen (Entrez) 1000
Especies diana Human
Contenido y almacenamiento Store at 4°C.
Formulario Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.