missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MYST1 Polyclonal antibody specifically detects MYST1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Aplicaciones | Western Blot, Immunohistochemistry |
| Clasificación | Polyclonal |
| Dilución | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Inmunógeno | The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human males absent on the first (MYST histone acetyltransferase 1) using the numbering given in entry BAB13924.1 (GeneID 84148). |
| ID de gen (Entrez) | 84148 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?