Learn More
MYST1 Antibody (7E11), Novus Biologicals™
Código de producto. 30026707 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μL
50 μL
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
30026707 100 μL 100 microlitros
30038719 50 μL 50 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 30026707 Proveedor Novus Biologicals N.º de proveedor NBP326184100ul
Rabbit Monoclonal Antibody
MYST1 Monoclonal antibody specifically detects MYST1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno MYST1
Aplicaciones ELISA, Immunohistochemistry, Immunocytochemistry
Clasificación Monoclonal
Clon 7E11
Conjugado Unconjugated
Dilución ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulación PBS, pH 7.4, 150mM NaCl and 50% glycerol
Alias de gen FLJ14040, hMOFhistone acetyltransferase MYST1, KAT8, MOFEC 2.3.1.48, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1, MYST histone acetyltransferase 1, MYST-1, ortholog of Drosophila males absent on the first (MOF), probable histone acetyltransferase MYST1
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from Human MYST1 [UniProt Q9H7Z6]
Método de purificación Affinity purified
Cantidad 100 μL
Estado normativo RUO
Disciplina de investigación Chromatin Research, DNA Repair
Primario o secundario Primary
ID de gen (Entrez) 84148
Especies diana Human
Contenido y almacenamiento Store at -20 to -70C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG