missing translation for 'onlineSavingsMsg'
Learn More

MYPN Antibody, Novus Biologicals™

Product Code. p-200074643 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
25 μL
Unit Size:
0.10 ml
25 microlitros
Product Code. Cantidad unitSize
18687185 25 μL 25 microlitros
18156418 0.1 mL 0.10 ml
2 options
This item is not returnable. View return policy

Product Code. 18687185

Brand: Novus Biologicals NBP23892425ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MYPN Polyclonal specifically detects MYPN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno MYPN
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulación PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
N.º de referencia del gen Q86TC9
Alias de gen 145 kDa (MYOP), MYOP145 kDa sarcomeric protein, myopalladin
Símbolos de los genes MYPN
Especie del huésped Rabbit
Inmunógeno This antibody was developed against a recombinant protein corresponding to amino acids: EDLSNNGSLHSANSTTNLAAIEPQPSPPHSEPPSVEQPPKPKLEGVLVNHNEPRSSSRIGLRVHFNLPEDDKGSEASSE
Método de purificación Affinity Purified
Cantidad 25 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 84665
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.