missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin heavy chain 11 Antibody (MYH11/2303R), DyLight 550, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Myosin heavy chain 11 Monoclonal antibody specifically detects Myosin heavy chain 11 in Human, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Rabbit samples. It is validated for Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | Myosin heavy chain 11 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | MYH11/2303R |
| Conjugado | DyLight 550 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant full-length human Myosin heavy chain 11 protein (Uniprot: P35749) |
| Método de purificación | Protein A or G purified |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?