Learn More
Novus Biologicals™ MyD88 Inhibitor Peptide Set
Código de producto. p-5538749 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
2 mg
Tamaño de la unidad:
2 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18766713 2 mg 2 miligramos
18759273 - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18766713 Proveedor Novus Biologicals™ N.º de proveedor NBP229328
Applications: Flow Cytometry, ELISA, Immunohistochemistry, In vitro assay, In vivo assay, Functional (Inhibition), Binding Inhibition, Block/Neutralize
TRUSTED_SUSTAINABILITY
Especificaciones
Contenido y almacenamiento Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Cantidad 2 mg
Para uso exclusivo en investigación