Learn More
Muscarinic Acetylcholine Receptor M2/CHRM2 Antibody (31-1D1), Novus Biologicals™
Código de producto. 18097174 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
125 μL
250 μL
Tamaño de la unidad:
125 microlitros
250 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18097174 125 μL 125 microlitros
18017465 250 μL 250 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18097174 Proveedor Novus Biologicals N.º de proveedor NB1202805125UL
Mouse Monoclonal Antibody
Muscarinic Acetylcholine Receptor M2/CHRM2 Monoclonal specifically detects Muscarinic Acetylcholine Receptor M2/CHRM2 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Muscarinic Acetylcholine Receptor M2/CHRM2
Aplicaciones Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Flow Cytometry, Immunoprecipitation, Western Blot
Clasificación Monoclonal
Clon 31-1D1
Concentración 2 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:1000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20
Formulación PBS with 0.05% Sodium Azide
Alias de gen 7TM receptor, cholinergic receptor, muscarinic 2, FLJ43243, HM2, MGC120006, MGC120007, muscarinic acetylcholine receptor M2, muscarinic M2 receptor
Símbolos de los genes CHRM2
Especie del huésped Mouse
Inmunógeno Purified porcine cardiac m2 muscarinic acetylcholine receptor.
Método de purificación Protein A purified
Cantidad 125 μL
Estado normativo RUO
Disciplina de investigación GPCR, Neuroscience
Primario o secundario Primary
ID de gen (Entrez) 1129
Especificidad de la prueba This is specific for the m2 mAChR subtype.
Especies diana Mouse, Pig, Human, Rat
Contenido y almacenamiento Store at -20C. Avoid freeze-thaw cycles.
Formulario Ascites
Isotype IgG1
Mostrar más Mostrar menos
For Research Use Only