missing translation for 'onlineSavingsMsg'
Learn More

MUC1 Antibody (MUC1/955), Alexa Fluor™ 350, Novus Biologicals™

Código de producto. 18133648 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18133648 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18133648 Proveedor Novus Biologicals N.º de proveedor NBP247884AF350

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MUC1 Monoclonal specifically detects MUC1 in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno MUC1
Aplicaciones Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Clasificación Monoclonal
Clon MUC1/955
Conjugado Alexa Fluor 350
Dilución Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
Formulación 50 mM sodium borate with 0.05% sodium azide
Alias de gen Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin
Símbolos de los genes MUC1
Especie del huésped Mouse
Inmunógeno Human milk-fat globule membranes (HMFGM) (Uniprot: P15941)
Método de purificación Protein A or G purified
Cantidad 0.1 mL
Disciplina de investigación Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 4582.0
Especificidad de la prueba This monoclonal antibody reacts with MUC1. The dominant epitope of this monoclonal antibody has not yet been determined. MUC1 is a large cell surface mucin glycoprotein expressed by most glandular and ductal epithelial cells and some hematopoietic cell lineages. It is expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. It is expressed abundantly in lactating mammary glands and over expressed abundantly in 90% breast carcinomas and metastases. Transgenic MUC1 has been shown to associate with all four c-erbB receptors and localize with c-erbB1 (EGFR) in lactating glands. The MUC1 gene contains seven exons and produces several different alternatively spliced variants. The major expressed form of MUC1 uses all seven exons and is a type 1 transmembrane protein with a large extracellular tandem repeat domain. The tandem repeat domain is highly O glycosylated and alterations in glycosylation have been shown in epithelial cancer cells. Antibody to EMA is useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in bone marrow or liver.
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4°C in the dark.
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.