missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
MUC1 Monoclonal antibody specifically detects MUC1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | MUC1 |
| Aplicaciones | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | MUC1/845 |
| Conjugado | DyLight 550 |
| Formulación | 50 mM Sodium Borate |
| Alias de gen | Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin |
| Especie del huésped | Mouse |
| Inmunógeno | Human milk-fat globule membranes (HMFGM) (Uniprot: P15941) |
| Método de purificación | Protein A or G purified |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?