missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
MTMR12 Polyclonal antibody specifically detects MTMR12 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | MTMR12 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | DyLight 405 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | 3-PAP3PAP, 3-phosphatase adapter subunit, KIAA1682FLJ20476, myotubularin related protein 12, myotubularin-related protein 12,3-phosphatase adapter protein, Phosphatidylinositol-3 phosphate 3-phosphatase adapter subunit, phosphatidylinositol-3 phosphate 3-phosphatase adaptor subunit, phosphatidylinositol-3-phosphate associated protein, PIP3AP |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with E. coli-derived Human MTMR12 fragment. |
| Método de purificación | Antigen and protein A Affinity-purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?