missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
MS4A4A Monoclonal antibody specifically detects MS4A4A in Human samples. It is validated for Flow Cytometry,CyTOF
Especificaciones
Especificaciones
| Antígeno | MS4A4A |
| Aplicaciones | Flow Cytometry, CyTOF |
| Clasificación | Monoclonal |
| Clon | 818112 |
| Conjugado | CoraFluor 1 |
| Formulación | PBS |
| Alias de gen | 4SPAN1, CD20 antigen-like 1, CD20L1CD20-L1, Fc epsilon receptor beta subunit homolog, Four-span transmembrane protein 1, membrane-spanning 4-domains subfamily A member 4A, membrane-spanning 4-domains, subfamily A, member 4, membrane-spanning 4-domains, subfamily A, member 4A, MS4A4MGC22311, MS4A7 |
| Especie del huésped | Mouse |
| Inmunógeno | Mouse myeloma cell line NS0 transfected with human MS4A4A, Accession # Q96JQ5 |
| Método de purificación | Protein A or G purified |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?