Learn More
MRPL17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA560458
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: PKLFQVLAPR YKDQTGGYTR MLQIPNRSLD RAKMAVIEYK GNCLPPLPLP RRDSHLTLLN QLLQGLRQDL RQSQEASNHS SHT Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 79%.
MRPL17 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include ribosome; structural constituent of ribosome; translation.Especificaciones
MRPL17 | |
Polyclonal | |
Unconjugated | |
MRPL17 | |
39S ribosomal protein L17, mitochondrial; L17mt; LIP2; LYST-interacting protein 2; LYST-interacting protein LIP2; Mitochondrial large ribosomal subunit protein bL17m; mitochondrial ribosomal protein L17; MRPL17; MRP-L17; MRP-L26; ribosomal protein, mitochondrial, L26; RPL17L; RPML26 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
63875 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NRX2 | |
MRPL17 | |
Recombinant protein corresponding to Human MRPL17 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only