missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse TRAIL (TNFSF10) Recombinant Protein, PeproTech®

Código de producto. 17802623
Change view
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
250 μg
500 μg
50 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
250 microgramos
50 microgramos
500 microgramos
Código de producto. Cantidad unitSize
17802623 500 μg 500 microgramos
17822013 100 μg 100 microgramos
17842333 10 μg 10 microgramos
17852913 1 mg 1 miligramo
17892823 250 μg 250 microgramos
17842823 50 μg 50 microgramos
6 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17802623

Marca: Gibco™ 31519500UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Murine TRAIL is a 174 amino acid polypeptide (20.0 kDa), consisting of the TNF-homologous portion of the extracellular domain of the full length TRAIL protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

TRAIL is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. TRAIL preferentially induces apoptosis in transformed and tumor cells, but does not appear to kill normal cells although it is expressed at a significant level in most normal tissues. TRAIL binds to several members of TNF receptor superfamily including TNFRSF10A/TRAILR1, TNFRSF10B/TRAILR2, TNFRSF10C/TRAILR3, TNFRSF10D/TRAILR4, and possibly also to TNFRSF11B/OPG. The activity of this protein may be modulated by binding to the decoy receptors TNFRSF10C/TRAILR3, TNFRSF10D/TRAILR4, and TNFRSF11B/OPG that cannot induce apoptosis. The binding of TRAIL protein to its receptors has been shown to trigger the activation of MAPK8/JNK, caspase 8, and caspase 3. Receptors for TRAIL include two death domain containing receptors, death receptor 4 (DR4) and DR5, as well as two decoy receptors, DcR1 and DcR2, lacking the intracellular signaling death domain. TRAIL is a type II membrane protein and expressed in a variety of human tissues. Like TNF and Fas ligand, TRAIL induces apoptosis and NF-kB activation in many tissues and cells.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P50592
Para utilizar con (aplicación) ELISA, Functional Assay, Western Blot
Formulación protein with no preservative
ID de gen (Entrez) 22035
Peso molecular 20 kDa
Nombre Mouse TRAIL (TNFSF10)
Cantidad 500 μg
Fuente E. coli
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen A330042I21Rik; AI448571; Apo-2 ligand; APO2L; Apo-2 L; CD253; CD253 antigen; chemokine tumor necrosis factor ligand superfamily member 10; Ly81; Protein TRAIL; TL2; TN; TNF superfamily member 10; TNF-related apoptosis inducing ligand; TNF-related apoptosis inducing ligand TRAIL; TNF-related apoptosis-inducing ligand; Tnfsf10; TNLG6A; TRAIL; TRAIL protein; TRAIL/APO2L; tumor necrosis factor (ligand) family, member 10; tumor necrosis factor (ligand) superfamily member 10; tumor necrosis factor (ligand) superfamily, member 10; tumor necrosis factor apoptosis-inducing ligand splice variant delta; tumor necrosis factor ligand 6 A; tumor necrosis factor ligand superfamily member 10; tumor necrosis factor superfamily member 10
Nombre común TRAIL (CD253)
Símbolo de gen TNFSF10
Actividad biológica Assay 1: Determined by the dose-dependent stimulation of MIP-2 production by mouse spleen cells using a concentration range of 10-100 ng/ml Assay 2: Measured by its ability to induce apoptosis in LN-18 cells (human glioblastoma cells). The expected ED50 for this effect is 40.0-60.0 ng/ml
Conjugado Unconjugated
Recombinante Recombinant
Secuencia MRGGRPQKVA AHITGITRRS NSALIPISKD GKTLGQKIES WESSRKGHSF LNHVLFRNGE LVIEQEGLYY IYSQTYFRFQ EAEDASKMVS KDKVRTKQLV QYIYKYTSYP DPIVLMKSAR NSCWSRDAEY GLYSIYQGGL FELKKNDRIF VSVTNEHLMD LDQEASFFGA FLIN
Contenido y almacenamiento -20°C
Sistema de expresión E. coli
Formulario Lyophilized
Grado de pureza o calidad ≥ 95% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.