missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Mouse TGF-Beta 1 / TGFB1 Protein

Código de producto. 17116821
Click to view available options
Cantidad:
1 mg
50 μg
Tamaño de la unidad:
1 miligramo
50 microgramos

Código de producto. 17116821

Marca: ACROBiosystems TG1M521850ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Transforming growth factor beta 1 ( TGFB1) is also known as TGF-β1, CED, DPD1, TGFB. is a polypeptide member of the transforming growth factor beta superfamily of cytokines. It is a secreted protein that performs many cellular functions, including the control of cell growth, cell proliferation, cell differentiation and apoptosis. The TGFB1 protein helps control the growth and division (proliferation) of cells, the process by which cells mature to carry out specific functions (differentiation), cell movement (motility), and the self-destruction of cells (apoptosis). The TGFB1 protein is found throughout the body and plays a role in development before birth, the formation of blood vessels, the regulation of muscle tissue and body fat development, wound healing, and immune system function. TGFB1 is particularly abundant in tissues that make up the skeleton, where it helps regulate bone growth, and in the intricate lattice that forms in the spaces between cells (the extracellular matrix). Within cells, this protein is turned off (inactive) until it receives a chemical signal to become active. TGFB1 plays an important role in controlling the immune system, and shows different activities on different types of cell, or cells at different developmental stages. Most immune cells (or leukocytes) secrete TGFB1. TGFB1 has been shown to interact with TGF beta receptor 1, LTBP1, YWHAE, EIF3I and Decorin.

This protein carries no "e;tag"e;. The protein has a calculated MW of 14.7 kDa.

Especificaciones

Peso molecular 14.7 kDa
Cantidad 50 μg
Fuente Mouse TGFB1 Protein (TG1-M5218) is expressed from human 293 cells (HEK293). It contains AA Ala 279 - Ser 390 (Accession # P04202-1).
Inmunógeno Ala 279 - Ser 390
Requisitos de almacenamiento -20°C
Alias de gen CEDLAP,TGF-beta 1,TGFB1,DPD1,TGF-beta-1,TGFB
Nombre común TGF-beta 1
Conjugado Unconjugated
Especie Mouse
Etiqueta de proteína Tag Free
Sistema de expresión HEK293
Formulario Powder
Subtipo de proteína Cytokines and Growth Factors
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado