missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Nogo Receptor/NgR Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 12 publications
Marca: R&D Systems AF1440
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Nogo Receptor/NgR Polyclonal specifically detects Nogo Receptor/NgR in Mouse samples. It is validated for Western Blot.
Especificaciones
| Nogo Receptor/NgR | |
| Polyclonal | |
| Unconjugated | |
| Q99PI8 | |
| RTN4R | |
| Mouse myeloma cell line NS0-derived recombinant mouse NgR Cys27-Ser447 Accession # Q99PI8 | |
| 100 μg | |
| Primary | |
| Detects mouse Nogo Receptor in direct ELISAs and Western blots. In these formats, approximately 10% cross-reactivity with recombinant human (rh) NgR is observed and less than 1% cross-reactivity with rhNgRH1 and rhNgRH2 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| NgR, NgR1, Nogo receptor, Nogo-66 receptor, NogoR, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor, RTN4R | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 65078 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido