missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse NGF Recombinant Protein

Código de producto. 17114293
Change view
Click to view available options
Cantidad:
50 μg
5 μg
Tamaño de la unidad:
5 microgramos
50 microgramos
Código de producto. Cantidad unitSize
17114293 50 μg 50 microgramos
17104293 5 μg 5 microgramos
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17114293

Marca: Invitrogen™ A42627

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Nerve growth factor (NGF) influences the survival and differentiation of a variety of neuronal and nonneuronal cells. The actions of this polypeptide result from binding to specific cell surface receptors which are present as both high and low affinity sites. Although both types of receptor bind NGF only the high affinity form is capable of signal transduction. Cross-linking studies with 125I-NGF have shown that the high affinity receptor of rat PC12 cells forms an Mr 158,000 complex with NGF while the low affinity receptor forms an Mr 100,000 complex. Full length cDNA clones for both the human and rat NGF receptor genes have been produced and sequenced, revealing greater than 90% homology between the two species. NGF exhibits nerve growth stimulating activity and is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons. Further, NGF stimulates division and differentiation of sympathetic and embryonic sensory neurons. Mutations in the NGF gene have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of NGF gene expression is associated with allergic rhinitis.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P01139
Para utilizar con (aplicación) Control, Bioactivity, Immunohistochemistry, Western Blot
Formulación 50mM sodium acetate with 5% mannitol, 150mM NaCl, 5% trehalose and no preservative; pH 5.5
ID de gen (Entrez) 18049
Peso molecular 13.5 kDa
Nombre Mouse NGF
Intervalo de pH 5.5
Método de purificación Purified
Cantidad 50 μg
Fuente CHO cells
Requisitos de almacenamiento -20°C or -80°C if preferred
Estado normativo RUO
Concentración de endotoxinas <1.0 EU/μg
Alias de gen B nerve growth factor; beta ngf; beta-nerve growth factor; BetaNGF; beta-NGF; bNGF; b-NGF; HSAN5; MGC161426; MGC161428; Nerve growth factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta polypeptide; nerve growth factor, beta subunit; NGF; NGF b; NGF β; Ngfb; NGFbeta; NGFβ; β nerve growth factor; β NGF; βNGF
Nombre común NGF
Símbolo de gen NGF
Actividad biológica Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. The ED50 for this effect is 1-6 ng/mL
Tipo de producto Protein
Conjugado Unconjugated
Especie Mouse
Recombinante Recombinant
Etiqueta de proteína Untagged
Secuencia Mouse NGF, amino acids Ser122-Gly241 (Accession # NP_001106168.1) with an initial C-terminal Met
Contenido y almacenamiento Store at -20 to -70 °C
Actividad Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. The ED50 for this effect is 1-6 ng/mL.
Nivel de endotoxinas < 1 EU/μg
Condiciones de envío Room Temperature
Sistema de expresión CHO cells
Familia de proteínas Growth Factors & Receptors
Forma de proteína Native
Formulario Lyophilized
Subtipo de proteína Neurotrophic Factors
Categoría de investigación Development, Immunology, Inflammation, Neurobiology
Grado de pureza o calidad >96% by SDS-PAGE
Expresión nerve growth factor
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.