missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse IL-33 Recombinant Protein, PeproTech®

Código de producto. 17820513
Change view
Click to view available options
Cantidad:
10 x 100 μg
100 μg
10 μg
2,5 x 100 μg
2 μg
5 x 100 μg
50 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
2 microgramos
250 microgramos
50 microgramos
500 microgramos
paquete de 10
paquete de 5
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17820513 10 μg 10 microgramos
17851283 100 μg 100 microgramos
30089567 10 x 100 μg paquete de 10
17860323 10 x 100 μg 1 miligramo
30089871 2,5 x 100 μg 250 microgramos
17869942 2,5 x 100 μg 250 microgramos
30089270 5 x 100 μg paquete de 5
17880323 2 μg 2 microgramos
17899942 5 x 100 μg 500 microgramos
17871283 50 μg 50 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17820513 Proveedor Gibco™ N.º de proveedor 2103310UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

210-33-1MG will be provided as 10 x 100 μg (210-33-100UG). 210-33-250UG will be provided as multiple 50 μg (210-33-50UG) and 100 μg (210-33-100UG) vials. 210-33-500UG will be provided as 5 x 100 μg (210-33-100UG). Recombinant Murine IL-33 is a 17.5 kDa protein containing 158 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-33 (Interleukin-33) is a 270 amino acid, highly divergent protein belonging to the IL-1 family with an IL-1-like C-terminal domain. IL-33 is a dual function protein that may function both as a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. IL-33 binds to and signals through IL1RL1/ST2 and its stimulation recruits MYD88, IRAK1, IRAK4, and TRAF6. IL-33 activates NF-kappaB and MAP kinases, and drives production of TH2-associated cytokines from in vitro polarized TH2 cells. In vivo, IL-33 induces the expression of IL-4, IL-5, and IL-13 and leads to severe pathological changes in mucosal organs. IL-33 is proteolytically converted to a mature form by CASP1 and is highly expressed in high endothelial venules found in tonsils, Peyer's patches and mesenteric lymph nodes and is almost undetectable in placenta. Prolonged IL-33 treatment of mice led to the development of eosinophilia, splenomegaly, and severe pathological changes in mucosal organs such as lungs, esophagus and small intestine. Recent experiments have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. Despite its predicted molecular weight, IL-33 will often run at higher molecular weight in SDS-PAGE. Studies have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine, and an intracellular nuclear factor with transcriptional regulatory properties.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P14719, Q8BVZ5
Para utilizar con (aplicación) Control, Bioactivity
Formulación Protein with no preservative
ID de gen (Entrez) 17082, 77125
Peso molecular 17.5 kDa
Nombre Mouse IL-33
Método de purificación Purified
Cantidad 10 μg
Fuente E. coli
Requisitos de almacenamiento -20°C
Estado normativo RUO
Concentración de endotoxinas <1 EU/μg
Alias de gen 9230117N10Rik; C9orf26; DVS27; DVS27-related protein; H-IL-33; IL1F11; IL-1F11; Il33; IL-33; ILN; Interleukin; interleukin 33; interleukin-1 family member 11; Interleukin33; Interleukin-33; Interleukin-33 (109-270); Interleukin-33 (95-270); Interleukin-33 (99-270); Interleukin-33(102-266); Interleukin-33(109-266); NFEHEV; NFHEV; NF-HEV; nuclear factor for high endothelial venules; nuclear factor from high endothelial venules; RGD1311155; RP11-575C20.2
Nombre común IL-33
Símbolo de gen IL1rl1, IL33
Actividad biológica The ED50 was determined by the dose-dependent stimulation of the proliferation of murine D10S cells is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg
Tipo de producto Protein
Conjugado Unconjugated
Recombinante Recombinant
Secuencia SIQGTSLLTQ SPASLSTYND QSVSFVLENG CYVINVDDSG KDQEQDQVLL RYYESPCPAS QSGDGVDGKK LMVNMSPIKD TDIWLHANDK DYSVELQRGD VSPPEQAFFV LHKKSSDFVS FECKNLPGTY IGVKDNQLAL VEEKDESCNN IMFKLSKI
Contenido y almacenamiento -20°C
Actividad The ED50 was determined by the dose-dependent stimulation of the proliferation of murine D10S cells is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg.
Sistema de expresión E. coli
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.